missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10588410
Click to view available options
Quantity:
500 g
1 kg
3 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum Potassium Sulfate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Weight (g/mol) 474.37
MDL Number 149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
IUPAC Name aluminium(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 92°C
Color White
pH 3.0 to 3.5
Physical Form Solid
Quantity 3 kg
Molecular Formula AlH24KO20S2
Formula Weight 474.39g/mol
Packaging Plastic powder jar
Product Identifier
  • Aluminium potassium sulfate dodecahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.