missing translation for 'onlineSavingsMsg'
Learn More

ACPT Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen ACPT
Concentration 0.27 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16882805
View Documents
Proteintech
24326-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16872805
View Documents
Proteintech
24326-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acid phosphatases are enzymes capable of hydrolyzing orthophosphoric acid esters in an acid medium. This gene is up-regulated by androgens and is down-regulated by estrogens in the prostate cancer cell line. This gene exhibits a lower level of expression in testicular cancer tissues than in normal tissues. The protein encoded by this gene has structural similarity to prostatic and lysosomal acid phosphatases. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications

Specifications

ACPT
Western Blot
Unconjugated
Rabbit
Mouse, Human
Q9BZG2
100503991, 93650
ACPT Fusion Protein Ag19283
Primary
-20°C
ACP4
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
acid phosphatase, testicular, ACPT, Testicular acid phosphatase
Acp4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.