missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACPT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24326-1-AP-150UL
This item is not returnable.
View return policy
Description
Acid phosphatases are enzymes capable of hydrolyzing orthophosphoric acid esters in an acid medium. This gene is up-regulated by androgens and is down-regulated by estrogens in the prostate cancer cell line. This gene exhibits a lower level of expression in testicular cancer tissues than in normal tissues. The protein encoded by this gene has structural similarity to prostatic and lysosomal acid phosphatases. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
ACPT | |
Polyclonal | |
Unconjugated | |
ACP4 | |
acid phosphatase, testicular, ACPT, Testicular acid phosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100503991, 93650 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BZG2 | |
Acp4 | |
ACPT Fusion Protein Ag19283 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction