missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, BAKER ULTRA RESI-ANALYZED, J.T. Baker™
Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Acetone, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.
127.00 € - 305.00 €
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10518521
|
JT Baker
9254.1000 |
1 L |
127.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10465651
|
JT Baker
9254.2500 |
2.5 L |
305.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.
- Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
- Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
- Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
- Performance is guaranteed through the expiration period stated on the label
- Suitable for exacting EPA sample cleanup protocols
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
5 to 6 (at 20°C) | |
67-64-1 | |
0.79g/cm3 | |
2.6 - 13 Vol % | |
465°C | |
Glass | |
Max. 1 ppm | |
0.3μeq/g | |
0.6μeq/g | |
32 mPas at 25°C | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
For Organic Residue Analysis |
-94.9°C | |
Colorless | |
55.8°C to 56.6°C | |
−20°C | |
Fruit-like | |
Liquid | |
C3H6O | |
(CH3)2CO | |
MFCD00008765 | |
Max. 0.5% | |
Fully miscible in water | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.4% | |
Acetone |
Safety and Handling
missing translation for 'einecsNumber' : 200-662-2
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.