missing translation for 'onlineSavingsMsg'
Learn More

Acetone, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 10465651
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Acetone, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 5 to 6 (at 20°C)
Melting Point -94.9°C
Color Colorless
Density 0.79g/cm3
Boiling Point 55.8°C to 56.6°C
Quantity 2.5 L
Explosion Limit 2.6 - 13 Vol %
Flash Point −20°C
Ignition Point 465°C
Odor Fruit-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Titratable Acid 0.3μeq/g
Linear Formula (CH3)2CO
Titratable Base 0.6μeq/g
Viscosity 32 mPas at 25°C
Water Max. 0.5%
Solubility Information Fully miscible in water
Formula Weight 58.08g/mol
Percent Purity ≥99.4%
Grade For Organic Residue Analysis
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'