missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, BAKER, J.T. Baker™
Meets reagent specifications for testing European Pharmacopeia monographs. For laboratory use, for organic synthesis, and for routine research.
159.00 € - 286.00 €
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10495871
|
JT Baker
8003.5000 |
5 L | EcoTainer Metal Safety Can |
159.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731472
|
JT Baker
8003.9010 |
10 L | JERRY |
286.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
5 to 6 (20°C) | |
67-64-1 | |
Passes test | |
56°C | |
−20°C | |
465°C | |
Liquid | |
C3H6O | |
MFCD00008765 | |
Max. 3g/L | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
Meets Ph. Eur. specifications, Manufactured in US FDA / ISO 9000 facility |
-94.7°C | |
Colorless | |
0.79g/mL at 20°C | |
2.6 - 12.8 Vol % | |
Passes test | |
Sweet, mint-like | |
Max. 50 ppm | |
(CH3)2CO | |
30.9 kPa (25°C) | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.0% | |
Acetone |
Safety and Handling
missing translation for 'einecsNumber' : 200-662-2
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title