missing translation for 'onlineSavingsMsg'
Learn More

Acetone, BAKER, J.T. Baker™

Product Code. 10731472
Click to view available options
Quantity:
5 L
10 L
Packaging:
EcoTainer Metal Safety Can
JERRY
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets reagent specifications for testing European Pharmacopeia monographs. For laboratory use, for organic synthesis, and for routine research.

A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH 5 to 6 (20°C)
Melting Point -94.7°C
Color Colorless
Appearance Passes test
Density 0.79g/mL at 20°C
Boiling Point 56°C
Quantity 10 L
Explosion Limit 2.6 - 12.8 Vol %
Flash Point −20°C
Identification Passes test
Ignition Point 465°C
Odor Sweet, mint-like
Packaging JERRY
Physical Form Liquid
Residue after Evaporation Max. 50 ppm
Linear Formula (CH3)2CO
Vapor Pressure 30.9 kPa (25°C)
Water Max. 3g/L
Formula Weight 58.08g/mol
Percent Purity ≥99.0%
Grade Meets Ph. Eur. specifications, Manufactured in US FDA / ISO 9000 facility
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.