missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetaldehyde di-n-hexyl acetal, 97%

15.00 €

Chemical Identifiers

CAS 5405-58-3
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.392
MDL Number MFCD00051715
InChI Key VSFOOCQEJQKBSO-UHFFFAOYSA-N
Synonym acetaldehyde, dihexyl acetal, acetaldehyde di-n-hexyl acetal, ethylidenebis oxy dihexane, acetaldehyde dihexylacetal, 1,1-di hexyloxy ethane, hexane, 1,1'-ethylidenebis oxy bis, acetaldehyde dihexyl acetal, 1-1-hexyloxy ethoxy hexane, 1,1-dihexyloxyethane, 1-1-hexoxyethoxy hexane
PubChem CID 21503
IUPAC Name 1-(1-hexoxyethoxy)hexane
SMILES CCCCCCOC(C)OCCCCCC
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460157
View Documents
Alfa Aesar™
B21064.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

5405-58-3
230.392
VSFOOCQEJQKBSO-UHFFFAOYSA-N
21503
CCCCCCOC(C)OCCCCCC
C14H30O2
MFCD00051715
acetaldehyde, dihexyl acetal, acetaldehyde di-n-hexyl acetal, ethylidenebis oxy dihexane, acetaldehyde dihexylacetal, 1,1-di hexyloxy ethane, hexane, 1,1'-ethylidenebis oxy bis, acetaldehyde dihexyl acetal, 1-1-hexyloxy ethoxy hexane, 1,1-dihexyloxyethane, 1-1-hexoxyethoxy hexane
1-(1-hexoxyethoxy)hexane

Specifications

5405-58-3
MFCD00051715
VSFOOCQEJQKBSO-UHFFFAOYSA-N
1-(1-hexoxyethoxy)hexane
21503
97%
C14H30O2
acetaldehyde, dihexyl acetal, acetaldehyde di-n-hexyl acetal, ethylidenebis oxy dihexane, acetaldehyde dihexylacetal, 1,1-di hexyloxy ethane, hexane, 1,1'-ethylidenebis oxy bis, acetaldehyde dihexyl acetal, 1-1-hexyloxy ethoxy hexane, 1,1-dihexyloxyethane, 1-1-hexoxyethoxy hexane
CCCCCCOC(C)OCCCCCC
230.392
230.39
Acetaldehyde di-n-hexyl acetal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 226-458-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.