missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetaldehyde di-n-hexyl acetal, 97%

Product Code. 11460157
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5405-58-3
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.392
MDL Number MFCD00051715
InChI Key VSFOOCQEJQKBSO-UHFFFAOYSA-N
Synonym acetaldehyde, dihexyl acetal, acetaldehyde di-n-hexyl acetal, ethylidenebis oxy dihexane, acetaldehyde dihexylacetal, 1,1-di hexyloxy ethane, hexane, 1,1'-ethylidenebis oxy bis, acetaldehyde dihexyl acetal, 1-1-hexyloxy ethoxy hexane, 1,1-dihexyloxyethane, 1-1-hexoxyethoxy hexane
PubChem CID 21503
IUPAC Name 1-(1-hexoxyethoxy)hexane
SMILES CCCCCCOC(C)OCCCCCC

Specifications

Quantity 5 g
Formula Weight 230.39
Percent Purity 97%
Chemical Name or Material Acetaldehyde di-n-hexyl acetal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.