missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one, 97%, Thermo Scientific™
74.10 € - 185.00 €
Chemical Identifiers
CAS | 50544-72-4 |
---|---|
Molecular Formula | C12H14O3 |
Molecular Weight (g/mol) | 206.241 |
MDL Number | MFCD01314121 |
InChI Key | OGVKPSHYVRAUCB-UHFFFAOYSA-N |
Synonym | 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-chromen-4-one, 7-hydroxy-2,2,8-trimethylchroman-4-one, 7-hydroxy-2,2,8-trimethyl-3h-1-benzopyran-4-one, 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-1-benzopyran-4-one, maybridge1_006374, 7-hydroxy-2,2,8-trimethyl4-chromanone, 7-hydroxy-2,2,8-trimethyl-4-chromanone, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-benzopyran-4-one, 4h-1-benzopyran-4-one,2,3-dihydro-7-hydroxy-2,2,8-trimethyl |
PubChem CID | 2727974 |
IUPAC Name | 7-hydroxy-2,2,8-trimethyl-3H-chromen-4-one |
SMILES | CC1=C(C=CC2=C1OC(CC2=O)(C)C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10681683
|
Thermo Scientific Maybridge
RH01364DA |
1 g | Amber Glass Bottle |
74.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10714013
|
Thermo Scientific Maybridge
RH01364EA |
10 g | Amber Glass Bottle |
185.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
50544-72-4 | |
206.241 | |
OGVKPSHYVRAUCB-UHFFFAOYSA-N | |
2727974 | |
CC1=C(C=CC2=C1OC(CC2=O)(C)C)O |
C12H14O3 | |
MFCD01314121 | |
7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-chromen-4-one, 7-hydroxy-2,2,8-trimethylchroman-4-one, 7-hydroxy-2,2,8-trimethyl-3h-1-benzopyran-4-one, 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-1-benzopyran-4-one, maybridge1_006374, 7-hydroxy-2,2,8-trimethyl4-chromanone, 7-hydroxy-2,2,8-trimethyl-4-chromanone, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-benzopyran-4-one, 4h-1-benzopyran-4-one,2,3-dihydro-7-hydroxy-2,2,8-trimethyl | |
7-hydroxy-2,2,8-trimethyl-3H-chromen-4-one |
Specifications
50544-72-4 | |
97% | |
MFCD01314121 | |
OGVKPSHYVRAUCB-UHFFFAOYSA-N | |
7-hydroxy-2,2,8-trimethyl-3H-chromen-4-one | |
2727974 | |
97% |
97.0 | |
C12H14O3 | |
7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-chromen-4-one, 7-hydroxy-2,2,8-trimethylchroman-4-one, 7-hydroxy-2,2,8-trimethyl-3h-1-benzopyran-4-one, 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-1-benzopyran-4-one, maybridge1_006374, 7-hydroxy-2,2,8-trimethyl4-chromanone, 7-hydroxy-2,2,8-trimethyl-4-chromanone, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-benzopyran-4-one, 4h-1-benzopyran-4-one,2,3-dihydro-7-hydroxy-2,2,8-trimethyl | |
CC1=C(C=CC2=C1OC(CC2=O)(C)C)O | |
206.241 | |
206.24 | |
7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one |