missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one, 97%, Thermo Scientific™

Product Code. 10714013
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50544-72-4
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01314121
InChI Key OGVKPSHYVRAUCB-UHFFFAOYSA-N
Synonym 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-chromen-4-one, 7-hydroxy-2,2,8-trimethylchroman-4-one, 7-hydroxy-2,2,8-trimethyl-3h-1-benzopyran-4-one, 7-hydroxy-2,2,8-trimethyl-2,3-dihydro-4h-1-benzopyran-4-one, maybridge1_006374, 7-hydroxy-2,2,8-trimethyl4-chromanone, 7-hydroxy-2,2,8-trimethyl-4-chromanone, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2,2,8-trimethyl-4h-benzopyran-4-one, 4h-1-benzopyran-4-one,2,3-dihydro-7-hydroxy-2,2,8-trimethyl
PubChem CID 2727974
IUPAC Name 7-hydroxy-2,2,8-trimethyl-3H-chromen-4-one
SMILES CC1=C(C=CC2=C1OC(CC2=O)(C)C)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 7-Hydroxy-2,2,8-trimethyl-2,3-dihydro-4H-chromen-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'