Learn More
6-Fluoroindole, 98%, Thermo Scientific Chemicals
63.10 € - 876.00 €
Chemical Identifiers
CAS | 399-51-9 |
---|---|
Molecular Formula | C8H6FN |
Molecular Weight (g/mol) | 135.14 |
MDL Number | MFCD00056933 |
InChI Key | YYFFEPUCAKVRJX-UHFFFAOYSA-N |
Synonym | 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g |
PubChem CID | 351278 |
IUPAC Name | 6-fluoro-1H-indole |
SMILES | FC1=CC=C2C=CNC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440498
|
Thermo Scientific Alfa Aesar
B24633.03 |
1 g |
63.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450498
|
Thermo Scientific Alfa Aesar
B24633.06 |
5 g |
214.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460498
|
Thermo Scientific Alfa Aesar
B24633.14 |
25 g |
876.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Chemical Identifiers
399-51-9 | |
135.14 | |
YYFFEPUCAKVRJX-UHFFFAOYSA-N | |
351278 | |
FC1=CC=C2C=CNC2=C1 |
C8H6FN | |
MFCD00056933 | |
6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g | |
6-fluoro-1H-indole |
Specifications
399-51-9 | |
C8H6FN | |
112192 | |
6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g | |
YYFFEPUCAKVRJX-UHFFFAOYSA-N | |
6-fluoro-1H-indole | |
351278 | |
98% |
74°C to 77°C | |
MFCD00056933 | |
Light sensitive | |
Insoluble in water. | |
FC1=CC=C2C=CNC2=C1 | |
135.14 | |
135.14 | |
6-Fluoroindole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only