missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoroindole, 98%, Thermo Scientific Chemicals

Product Code. 11450498
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoroindole acts as a reagent in the synthesis of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles, which acts as a potential anticancer immunomodulator. It is also employed as an antifungal and antibacterial agent. Further, it serves as a potent selective serotonin reuptake inhibitor. In addition to this, it is used as a inhibitor of HIV-1 attachment.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool place.Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-51-9
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00056933
InChI Key YYFFEPUCAKVRJX-UHFFFAOYSA-N
Synonym 6-fluoroindole, 1h-indole, 6-fluoro, 6-fluoro indole, 6-fluoroindol, 6-fluoro-indole, pubchem1682, acmc-1cty0, ksc222e4t, 6-fluoroindole 1g
PubChem CID 351278
IUPAC Name 6-fluoro-1H-indole
SMILES FC1=CC=C2C=CNC2=C1

Specifications

Melting Point 74°C to 77°C
Quantity 5 g
Beilstein 112192
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 135.14
Percent Purity 98%
Chemical Name or Material 6-Fluoroindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'