Learn More
5-Methylisatin, 97%, Thermo Scientific Chemicals
34.57 € - 418.08 €
Chemical Identifiers
CAS | 608-05-9 |
---|---|
Molecular Formula | C9H7NO2 |
Molecular Weight (g/mol) | 161.16 |
MDL Number | MFCD00005721 |
InChI Key | VAJCSPZKMVQIAP-UHFFFAOYSA-N |
Synonym | 5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione |
PubChem CID | 11840 |
IUPAC Name | 5-methyl-1H-indole-2,3-dione |
SMILES | CC1=CC=C2NC(=O)C(=O)C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367935
|
Thermo Scientific Alfa Aesar
L08009.03 |
1 g |
34.57 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377935
|
Thermo Scientific Alfa Aesar
L08009.06 |
5 g |
114.57 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387935
|
Thermo Scientific Alfa Aesar
L08009.14 |
25 g |
418.08 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
608-05-9 | |
161.16 | |
VAJCSPZKMVQIAP-UHFFFAOYSA-N | |
11840 | |
CC1=CC=C2NC(=O)C(=O)C2=C1 |
C9H7NO2 | |
MFCD00005721 | |
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione | |
5-methyl-1H-indole-2,3-dione |
Specifications
608-05-9 | |
C9H7NO2 | |
123738 | |
VAJCSPZKMVQIAP-UHFFFAOYSA-N | |
5-methyl-1H-indole-2,3-dione | |
11840 | |
97% |
∽180°C (decomposition) | |
MFCD00005721 | |
5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione | |
CC1=CC=C2NC(=O)C(=O)C2=C1 | |
161.16 | |
161.16 | |
5-Methylisatin |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-152-1
missing translation for 'rtecsNumber' : NL7939300
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.