missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisatin, 97%, Thermo Scientific Chemicals

Product Code. 11377935
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-05-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005721
InChI Key VAJCSPZKMVQIAP-UHFFFAOYSA-N
Synonym 5-methylisatin, 5-methylindoline-2,3-dione, 5-methylindole-2,3-dione, 1h-indole-2,3-dione, 5-methyl, indole-2,3-dione, 5-methyl, 5-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-methyl-2,3-indolinedione, 5-methyl-indole-2,3-dione, 5-methylindole-2,3 1h-dione, 5-methyl-1h-benzo d azolidine-2,3-dione
PubChem CID 11840
IUPAC Name 5-methyl-1H-indole-2,3-dione
SMILES CC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point ∽180°C (decomposition)
Quantity 5 g
Beilstein 123738
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 5-Methylisatin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'