Learn More
5-Fluoropyridine-3-boronic acid, 98%, Thermo Scientific Chemicals
78.00 € - 234.00 €
Chemical Identifiers
CAS | 872041-86-6 |
---|---|
Molecular Formula | C5H5BFNO2 |
Molecular Weight (g/mol) | 140.908 |
MDL Number | MFCD07368243 |
InChI Key | FVEDGBRHTGXPOK-UHFFFAOYSA-N |
Synonym | 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine |
PubChem CID | 44717403 |
IUPAC Name | (5-fluoropyridin-3-yl)boronic acid |
SMILES | B(C1=CC(=CN=C1)F)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489627
|
Thermo Scientific Alfa Aesar
H52860.MD |
250 mg |
78.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479627
|
Thermo Scientific Alfa Aesar
H52860.03 |
1 g |
234.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.
Solubility
Soluble in hot methanol.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
872041-86-6 | |
140.908 | |
FVEDGBRHTGXPOK-UHFFFAOYSA-N | |
44717403 | |
B(C1=CC(=CN=C1)F)(O)O |
C5H5BFNO2 | |
MFCD07368243 | |
5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine | |
(5-fluoropyridin-3-yl)boronic acid |
Specifications
872041-86-6 | |
C5H5BFNO2 | |
5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine | |
FVEDGBRHTGXPOK-UHFFFAOYSA-N | |
(5-fluoropyridin-3-yl)boronic acid | |
44717403 | |
98% |
233°C to 236°C | |
MFCD07368243 | |
Soluble in hot methanol. | |
B(C1=CC(=CN=C1)F)(O)O | |
140.908 | |
140.91 | |
5-Fluoropyridine-3-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.