missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoropyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15479627
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoropyridine-3-boronic acid is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872041-86-6
Molecular Formula C5H5BFNO2
Molecular Weight (g/mol) 140.908
MDL Number MFCD07368243
InChI Key FVEDGBRHTGXPOK-UHFFFAOYSA-N
Synonym 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine
PubChem CID 44717403
IUPAC Name (5-fluoropyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1)F)(O)O

Specifications

Melting Point 233°C to 236°C
Quantity 1 g
Solubility Information Soluble in hot methanol.
Formula Weight 140.91
Percent Purity 98%
Chemical Name or Material 5-Fluoropyridine-3-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.