missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloroisatin, 98%, Thermo Scientific Chemicals
21.00 € - 96.88 €
Chemical Identifiers
CAS | 17630-76-1 |
---|---|
Molecular Formula | C8H4ClNO2 |
Molecular Weight (g/mol) | 181.58 |
MDL Number | MFCD00014567 |
InChI Key | XHDJYQWGFIBCEP-UHFFFAOYSA-N |
Synonym | 5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607 |
PubChem CID | 87203 |
SMILES | ClC1=CC=C2NC(=O)C(=O)C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484474
|
Thermo Scientific Alfa Aesar
A13125.06 |
5 g |
21.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494474
|
Thermo Scientific Alfa Aesar
A13125.14 |
25 g |
31.56 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404484
|
Thermo Scientific Alfa Aesar
A13125.22 |
100 g |
96.88 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
17630-76-1 | |
181.58 | |
XHDJYQWGFIBCEP-UHFFFAOYSA-N | |
87203 |
C8H4ClNO2 | |
MFCD00014567 | |
5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607 | |
ClC1=CC=C2NC(=O)C(=O)C2=C1 |
Specifications
17630-76-1 | |
C8H4ClNO2 | |
383759 | |
It is insoluble in water. | |
ClC1=CC=C2NC(=O)C(=O)C2=C1 | |
181.58 | |
181.58 | |
5-Chloroisatin |
∼247°C (decomposition) | |
MFCD00014567 | |
5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607 | |
XHDJYQWGFIBCEP-UHFFFAOYSA-N | |
5-chloro-2,3-dihydro-1H-indole-2,3-dione | |
87203 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 241-614-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only