missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroisatin, 98%, Thermo Scientific Chemicals

Product Code. 11494474
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17630-76-1
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.58
MDL Number MFCD00014567
InChI Key XHDJYQWGFIBCEP-UHFFFAOYSA-N
Synonym 5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607
PubChem CID 87203
SMILES ClC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point ∼247°C (decomposition)
Quantity 25 g
Beilstein 383759
Solubility Information It is insoluble in water.
IUPAC Name 5-chloro-1H-indole-2,3-dione
Formula Weight 181.58
Percent Purity 98%
Chemical Name or Material 5-Chloroisatin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'