Learn More
5-Chloroindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals
54.30 € - 612.00 €
Chemical Identifiers
CAS | 10517-21-2 |
---|---|
Molecular Formula | C9H5ClNO2 |
Molecular Weight (g/mol) | 194.59 |
MDL Number | MFCD00005613 |
InChI Key | FUQOTYRCMBZFOL-UHFFFAOYSA-M |
Synonym | 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 |
PubChem CID | 82693 |
SMILES | [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 |
Description
Potent antagonist at the glycine site of the NMDA receptor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10517-21-2 | |
194.59 | |
FUQOTYRCMBZFOL-UHFFFAOYSA-M | |
82693 |
C9H5ClNO2 | |
MFCD00005613 | |
5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 | |
[O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 |
Specifications
10517-21-2 | |
C9H5ClNO2 | |
153229 | |
FUQOTYRCMBZFOL-UHFFFAOYSA-M | |
5-chloro-1H-indole-2-carboxylate | |
82693 | |
98% |
287°C (decomposition) | |
MFCD00005613 | |
5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783 | |
[O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1 | |
194.59 | |
195.6 | |
5-Chloroindole-2-carboxylic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 234-050-1
missing translation for 'rtecsNumber' : NL5998400
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.