missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11458567
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent antagonist at the glycine site of the NMDA receptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10517-21-2
Molecular Formula C9H5ClNO2
Molecular Weight (g/mol) 194.59
MDL Number MFCD00005613
InChI Key FUQOTYRCMBZFOL-UHFFFAOYSA-M
Synonym 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783
PubChem CID 82693
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1

Specifications

Melting Point 287°C (decomposition)
Quantity 5 g
Beilstein 153229
IUPAC Name 5-chloro-1H-indole-2-carboxylic acid
Formula Weight 195.6
Percent Purity 98%
Chemical Name or Material 5-Chloroindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'