missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzonitrile, 97%, Thermo Scientific™

27.70 € - 93.60 €

Chemical Identifiers

CAS 40530-18-5
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00143428
InChI Key PVCONXMDUZOPJH-UHFFFAOYSA-N
Synonym 2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromosalicylonitrile, 5-bromo-2-hydroxy-benzonitrile, buttpark 20\02-76, benzonitrile, 5-bromo-2-hydroxy, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, pubchem3782, acmc-1apuz
PubChem CID 2757014
IUPAC Name 5-bromo-2-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471548
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55333.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15481548
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55333.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

40530-18-5
198.019
PVCONXMDUZOPJH-UHFFFAOYSA-N
2757014
C1=CC(=C(C=C1Br)C#N)O
C7H4BrNO
MFCD00143428
2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromosalicylonitrile, 5-bromo-2-hydroxy-benzonitrile, buttpark 20\02-76, benzonitrile, 5-bromo-2-hydroxy, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, pubchem3782, acmc-1apuz
5-bromo-2-hydroxybenzonitrile

Specifications

40530-18-5
C7H4BrNO
2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromosalicylonitrile, 5-bromo-2-hydroxy-benzonitrile, buttpark 20\02-76, benzonitrile, 5-bromo-2-hydroxy, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, pubchem3782, acmc-1apuz
C1=CC(=C(C=C1Br)C#N)O
198.019
198.02
5-Bromo-2-hydroxybenzonitrile
158°C to 163°C
MFCD00143428
PVCONXMDUZOPJH-UHFFFAOYSA-N
5-bromo-2-hydroxybenzonitrile
2757014
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 254-958-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications