missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-hydroxybenzonitrile, 97%, Thermo Scientific™

Product Code. 15481548
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40530-18-5
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD00143428
InChI Key PVCONXMDUZOPJH-UHFFFAOYSA-N
Synonym 2-hydroxy-5-bromobenzonitrile, 4-bromo-2-cyanophenol, 5-bromosalicylonitrile, 5-bromo-2-hydroxy-benzonitrile, buttpark 20\02-76, benzonitrile, 5-bromo-2-hydroxy, 5-bromo-2-hydroxybenzenecarbonitrile, 5-bromosalicylnitrile, pubchem3782, acmc-1apuz
PubChem CID 2757014
IUPAC Name 5-bromo-2-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1Br)C#N)O

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Formula Weight 198.02
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxybenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'