Learn More
5-Benzoylpentanoic acid, 99%, Thermo Scientific Chemicals
25.20 € - 252.00 €
Chemical Identifiers
CAS | 4144-62-1 |
---|---|
Molecular Formula | C12H14O3 |
Molecular Weight (g/mol) | 206.24 |
MDL Number | MFCD00014380 |
InChI Key | AIEMSTCGCMIJTI-UHFFFAOYSA-N |
Synonym | 5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid |
PubChem CID | 223595 |
IUPAC Name | 6-oxo-6-phenylhexanoic acid |
SMILES | OC(=O)CCCCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318085
|
Thermo Scientific Alfa Aesar
L08290.03 |
1 g |
25.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328085
|
Thermo Scientific Alfa Aesar
L08290.06 |
5 g |
72.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338085
|
Thermo Scientific Alfa Aesar
L08290.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4144-62-1 | |
206.24 | |
AIEMSTCGCMIJTI-UHFFFAOYSA-N | |
223595 | |
OC(=O)CCCCC(=O)C1=CC=CC=C1 |
C12H14O3 | |
MFCD00014380 | |
5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid | |
6-oxo-6-phenylhexanoic acid |
Specifications
4144-62-1 | |
C12H14O3 | |
1961325 | |
AIEMSTCGCMIJTI-UHFFFAOYSA-N | |
6-oxo-6-phenylhexanoic acid | |
223595 | |
99% |
75°C to 79°C | |
MFCD00014380 | |
5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid | |
OC(=O)CCCCC(=O)C1=CC=CC=C1 | |
206.24 | |
206.24 | |
5-Benzoylpentanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only