missing translation for 'onlineSavingsMsg'
Learn More

5-Benzoylpentanoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11338085
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4144-62-1
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00014380
InChI Key AIEMSTCGCMIJTI-UHFFFAOYSA-N
Synonym 5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid
PubChem CID 223595
IUPAC Name 6-oxo-6-phenylhexanoic acid
SMILES OC(=O)CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 75°C to 79°C
Quantity 25 g
Beilstein 1961325
Formula Weight 206.24
Percent Purity 99%
Chemical Name or Material 5-Benzoylpentanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.