missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-n-Nonylacetophenone, 97%, Thermo Scientific™
18.10 € - 68.80 €
Chemical Identifiers
CAS | 37593-05-8 |
---|---|
Molecular Formula | C17H26O |
Molecular Weight (g/mol) | 246.394 |
MDL Number | MFCD00043717 |
InChI Key | PWRSUUUQEQAMEV-UHFFFAOYSA-N |
Synonym | p-nonylacetophenone, 1-4-nonylphenyl ethan-1-one, 4-n-nonylacetophenone, 1-4-nonylphenyl ethanone, 4'-n-nonylacetophenone, p-nonyl acetophenone, 4'-nonylacetophenone, 1-acetyl-4-nonylbenzene |
PubChem CID | 142175 |
IUPAC Name | 1-(4-nonylphenyl)ethanone |
SMILES | CCCCCCCCCC1=CC=C(C=C1)C(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11455223
|
Thermo Scientific Alfa Aesar
A12831.03 |
1 g | |||||||
11465223
|
Thermo Scientific Alfa Aesar
A12831.06 |
5 g |
68.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
37593-05-8 | |
246.394 | |
PWRSUUUQEQAMEV-UHFFFAOYSA-N | |
142175 | |
CCCCCCCCCC1=CC=C(C=C1)C(=O)C |
C17H26O | |
MFCD00043717 | |
p-nonylacetophenone, 1-4-nonylphenyl ethan-1-one, 4-n-nonylacetophenone, 1-4-nonylphenyl ethanone, 4'-n-nonylacetophenone, p-nonyl acetophenone, 4'-nonylacetophenone, 1-acetyl-4-nonylbenzene | |
1-(4-nonylphenyl)ethanone |
Specifications
37593-05-8 | |
C17H26O | |
MFCD00043717 | |
PWRSUUUQEQAMEV-UHFFFAOYSA-N | |
1-(4-nonylphenyl)ethanone | |
142175 | |
97% |
159°C to 161°C (2 mmHg) | |
1.505 | |
p-nonylacetophenone, 1-4-nonylphenyl ethan-1-one, 4-n-nonylacetophenone, 1-4-nonylphenyl ethanone, 4'-n-nonylacetophenone, p-nonyl acetophenone, 4'-nonylacetophenone, 1-acetyl-4-nonylbenzene | |
CCCCCCCCCC1=CC=C(C=C1)C(=O)C | |
246.394 | |
246.39 | |
4'-n-Nonylacetophenone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures