missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Nonylacetophenone, 97%, Thermo Scientific™

Product Code. 11465223
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37593-05-8
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.394
MDL Number MFCD00043717
InChI Key PWRSUUUQEQAMEV-UHFFFAOYSA-N
Synonym p-nonylacetophenone, 1-4-nonylphenyl ethan-1-one, 4-n-nonylacetophenone, 1-4-nonylphenyl ethanone, 4'-n-nonylacetophenone, p-nonyl acetophenone, 4'-nonylacetophenone, 1-acetyl-4-nonylbenzene
PubChem CID 142175
IUPAC Name 1-(4-nonylphenyl)ethanone
SMILES CCCCCCCCCC1=CC=C(C=C1)C(=O)C

Specifications

Boiling Point 159°C to 161°C (2 mmHg)
Refractive Index 1.505
Quantity 5 g
Formula Weight 246.39
Percent Purity 97%
Chemical Name or Material 4'-n-Nonylacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'