missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(N-Boc-methylamino)benzoic acid, 97%, Thermo Scientific™
40.00 € - 461.00 €
Chemical Identifiers
CAS | 263021-30-3 |
---|---|
Molecular Formula | C13H17NO4 |
Molecular Weight (g/mol) | 251.28 |
MDL Number | MFCD04972273 |
InChI Key | VRQKKKZQZSNRKD-UHFFFAOYSA-N |
Synonym | 4-tert-butoxycarbonyl methyl amino benzoic acid, n-boc-4-methylamino benzoic acid, methyl amino benzoic acid, 4-tert-butoxycarbonyl, 4-n-boc-methylamino benzoic acid, 4-n-boc-n-methylamino benzoic acid, n-boc-4-methylamino benzoicacid, 4-tert-butoxycarbonyl-methyl-amino-benzoic acid |
PubChem CID | 11172762 |
IUPAC Name | 4-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]benzoic acid |
SMILES | CN(C(=O)OC(C)(C)C)C1=CC=C(C=C1)C(O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-(N-Boc-methylamino)benzoic acid is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
263021-30-3 | |
251.28 | |
VRQKKKZQZSNRKD-UHFFFAOYSA-N | |
11172762 | |
CN(C(=O)OC(C)(C)C)C1=CC=C(C=C1)C(O)=O |
C13H17NO4 | |
MFCD04972273 | |
4-tert-butoxycarbonyl methyl amino benzoic acid, n-boc-4-methylamino benzoic acid, methyl amino benzoic acid, 4-tert-butoxycarbonyl, 4-n-boc-methylamino benzoic acid, 4-n-boc-n-methylamino benzoic acid, n-boc-4-methylamino benzoicacid, 4-tert-butoxycarbonyl-methyl-amino-benzoic acid | |
4-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]benzoic acid |
Specifications
263021-30-3 | |
MFCD04972273 | |
VRQKKKZQZSNRKD-UHFFFAOYSA-N | |
4-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]benzoic acid | |
11172762 | |
97% |
C13H17NO4 | |
4-tert-butoxycarbonyl methyl amino benzoic acid, n-boc-4-methylamino benzoic acid, methyl amino benzoic acid, 4-tert-butoxycarbonyl, 4-n-boc-methylamino benzoic acid, 4-n-boc-n-methylamino benzoic acid, n-boc-4-methylamino benzoicacid, 4-tert-butoxycarbonyl-methyl-amino-benzoic acid | |
CN(C(=O)OC(C)(C)C)C1=CC=C(C=C1)C(O)=O | |
251.28 | |
251.28 | |
4-(N-Boc-methylamino)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures