missing translation for 'onlineSavingsMsg'
Learn More

4-(N-Boc-methylamino)benzoic acid, 97%, Thermo Scientific™

Product Code. 15449297
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(N-Boc-methylamino)benzoic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 263021-30-3
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD04972273
InChI Key VRQKKKZQZSNRKD-UHFFFAOYSA-N
Synonym 4-tert-butoxycarbonyl methyl amino benzoic acid, n-boc-4-methylamino benzoic acid, methyl amino benzoic acid, 4-tert-butoxycarbonyl, 4-n-boc-methylamino benzoic acid, 4-n-boc-n-methylamino benzoic acid, n-boc-4-methylamino benzoicacid, 4-tert-butoxycarbonyl-methyl-amino-benzoic acid
PubChem CID 11172762
IUPAC Name 4-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]benzoic acid
SMILES CN(C(=O)OC(C)(C)C)C1=CC=C(C=C1)C(O)=O

Specifications

Quantity 5 g
Formula Weight 251.28
Percent Purity 97%
Chemical Name or Material 4-(N-Boc-methylamino)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'