missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylthio)butanol, 99%, Thermo Scientific Chemicals
31.90 € - 328.00 €
Chemische Identifikatoren
CAS | 20582-85-8 |
---|---|
Molecular Formula | C5H12OS |
Molecular Weight (g/mol) | 120.21 |
MDL Number | MFCD00002973 |
InChI Key | JNTVUHZXIJFHAU-UHFFFAOYSA-N |
Synonym | 4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound |
PubChem CID | 519793 |
SMILES | CSCCCCO |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11465307
|
Thermo Scientific Alfa Aesar
B22673.03 |
1 g |
31.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475307
|
Thermo Scientific Alfa Aesar
B22673.06 |
5 g |
97.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485307
|
Thermo Scientific Alfa Aesar
B22673.14 |
25 g |
328.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
20582-85-8 | |
120.21 | |
JNTVUHZXIJFHAU-UHFFFAOYSA-N | |
519793 |
C5H12OS | |
MFCD00002973 | |
4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound | |
CSCCCCO |
Spezifikation
20582-85-8 | |
95°C to 98°C (20 mmHg) | |
Unpleasant | |
CH3S(CH2)4OH | |
MFCD00002973 | |
JNTVUHZXIJFHAU-UHFFFAOYSA-N | |
4-methylsulfanylbutan-1-ol | |
519793 | |
99% |
0.999 | |
101°C (215°F) | |
C5H12OS | |
1.487 | |
4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound | |
CSCCCCO | |
120.21 | |
120.21 | |
4-(Methylthio)butanol |
Sicherheit und Handhabung
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only