missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)butanol, 99%, Thermo Scientific Chemicals

Product Code. 11475307
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20582-85-8
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00002973
InChI Key JNTVUHZXIJFHAU-UHFFFAOYSA-N
Synonym 4-methylthio-1-butanol, 4-methylthio butanol, 4-methylthiobutan-1-ol, unii-yl87ury6pc, 4-methylsulfanyl-1-butanol, 1-butanol, 4-methylthio, yl87ury6pc, 4-methylthio butan-1-ol, 4-methylsulfanyl butan-1-ol, 4-methylthio-1-butanol, polymer-bound
PubChem CID 519793
SMILES CSCCCCO

Specifications

Density 0.999
Boiling Point 95°C to 98°C (20 mmHg)
Flash Point 101°C (215°F)
Odor Unpleasant
Linear Formula CH3S(CH2)4OH
Refractive Index 1.487
Quantity 5 g
IUPAC Name 4-(methylsulfanyl)butan-1-ol
Formula Weight 120.21
Percent Purity 99%
Chemical Name or Material 4-(Methylthio)butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'