missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals

34.57 € - 381.90 €

Chemical Identifiers

CAS 13329-40-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00045320
InChI Key JZJWCDQGIPQBAO-UHFFFAOYSA-N
Synonym 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
PubChem CID 72869
IUPAC Name 1-(4-iodophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11492818
View Documents
Thermo Scientific Alfa Aesar
A16794.06
5 g
34.57 €
5g
Please to purchase this item. Need a web account? Register with us today!
11402828
View Documents
Thermo Scientific Alfa Aesar
A16794.14
25 g
114.57 €
25g
Please to purchase this item. Need a web account? Register with us today!
11412828
View Documents
Thermo Scientific Alfa Aesar
A16794.22
100 g
381.90 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

13329-40-3
246.047
JZJWCDQGIPQBAO-UHFFFAOYSA-N
72869
CC(=O)C1=CC=C(C=C1)I
C8H7IO
MFCD00045320
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
1-(4-iodophenyl)ethanone

Specifications

13329-40-3
C8H7IO
1857412
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
JZJWCDQGIPQBAO-UHFFFAOYSA-N
1-(4-iodophenyl)ethanone
72869
98%
82°C to 84°C
MFCD00045320
Light sensitive
Easily soluble in water.
CC(=O)C1=CC=C(C=C1)I
246.047
246.05
4'-Iodoacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 236-372-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.