missing translation for 'onlineSavingsMsg'
Learn More

4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11402828
Click to view available options
Quantity:
5 g
25 g
100 g

Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions

Solubility
Easily soluble in water.

Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13329-40-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.047
MDL Number MFCD00045320
InChI Key JZJWCDQGIPQBAO-UHFFFAOYSA-N
Synonym 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone
PubChem CID 72869
IUPAC Name 1-(4-iodophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)I

Specifications

Melting Point 82°C to 84°C
Quantity 25 g
Beilstein 1857412
Sensitivity Light sensitive
Solubility Information Easily soluble in water.
Formula Weight 246.05
Percent Purity 98%
Chemical Name or Material 4'-Iodoacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'