missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N,N-dimethyl-2-nitroaniline, 97%, Thermo Scientific™

27.00 € - 288.00 €

Chemical Identifiers

CAS 69261-01-4
Molecular Formula C8H9FN2O2
Molecular Weight (g/mol) 184.17
MDL Number MFCD04116334
InChI Key RGCUVPLVTHHXMT-UHFFFAOYSA-N
Synonym n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
PubChem CID 16440018
IUPAC Name 4-fluoro-N,N-dimethyl-2-nitroaniline
SMILES CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15490009
View Documents
Thermo Scientific Alfa Aesar
H63357.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15470009
View Documents
Thermo Scientific Alfa Aesar
H63357.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15480009
View Documents
Thermo Scientific Alfa Aesar
H63357.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

69261-01-4
184.17
RGCUVPLVTHHXMT-UHFFFAOYSA-N
16440018
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
C8H9FN2O2
MFCD04116334
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
4-fluoro-N,N-dimethyl-2-nitroaniline

Specifications

69261-01-4
MFCD04116334
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
184.17
184.17
Solid
C8H9FN2O2
UN2811
RGCUVPLVTHHXMT-UHFFFAOYSA-N
4-fluoro-N,N-dimethyl-2-nitroaniline
16440018
97%
4-Fluoro-N,N-dimethyl-2-nitroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications