missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N,N-dimethyl-2-nitroaniline, 97%, Thermo Scientific™

Product Code. 15470009
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69261-01-4
Molecular Formula C8H9FN2O2
Molecular Weight (g/mol) 184.17
MDL Number MFCD04116334
InChI Key RGCUVPLVTHHXMT-UHFFFAOYSA-N
Synonym n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
PubChem CID 16440018
IUPAC Name 4-fluoro-N,N-dimethyl-2-nitroaniline
SMILES CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 184.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-N,N-dimethyl-2-nitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'