Learn More
4-Chloro-7-azaindole, 98%, Thermo Scientific Chemicals
67.60 € - 749.00 €
Chemical Identifiers
CAS | 55052-28-3 |
---|---|
Molecular Formula | C7H5ClN2 |
Molecular Weight (g/mol) | 152.58 |
MDL Number | MFCD08272232 |
InChI Key | HNTZVGMWXCFCTA-UHFFFAOYSA-N |
Synonym | 4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine |
PubChem CID | 11389493 |
IUPAC Name | 4-chloro-1H-pyrrolo[2,3-b]pyridine |
SMILES | ClC1=C2C=CNC2=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15483268
|
Thermo Scientific Alfa Aesar
H56734.MD |
250 mg |
67.60 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15463268
|
Thermo Scientific Alfa Aesar
H56734.03 |
1 g |
180.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473268
|
Thermo Scientific Alfa Aesar
H56734.06 |
5 g | N/A | ||||||
Description
A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
55052-28-3 | |
152.58 | |
HNTZVGMWXCFCTA-UHFFFAOYSA-N | |
11389493 | |
ClC1=C2C=CNC2=NC=C1 |
C7H5ClN2 | |
MFCD08272232 | |
4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine | |
4-chloro-1H-pyrrolo[2,3-b]pyridine |
Specifications
55052-28-3 | |
C7H5ClN2 | |
UN2811 | |
Slightly soluble in water. | |
ClC1=C2C=CNC2=NC=C1 | |
152.58 | |
152.58 | |
4-Chloro-7-azaindole |
176°C to 181°C | |
MFCD08272232 | |
4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine | |
HNTZVGMWXCFCTA-UHFFFAOYSA-N | |
4-chloro-1H-pyrrolo[2,3-b]pyridine | |
11389493 | |
98% |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.