missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-azaindole, 98%, Thermo Scientific Chemicals

Product Code. 15463268
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful intermediate for drug discovery research such as used in the synthesis of 7-azaindole derivatives, synthetic cytokinin analogues.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55052-28-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD08272232
InChI Key HNTZVGMWXCFCTA-UHFFFAOYSA-N
Synonym 4-chloro-7-azaindole, 4-chloro-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 4-chloro, 4-choro-7-azaindole, pubchem17277, acmc-1ay8o, 4-chloro-7-azaindol, ksc494a8l, 4-chloropyrrolo 2,3-b pyridine
PubChem CID 11389493
IUPAC Name 4-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES ClC1=C2C=CNC2=NC=C1

Specifications

Melting Point 176°C to 181°C
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 152.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-7-azaindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'