Learn More
4-Bromo-2-fluorophenol, 97+%, Thermo Scientific Chemicals
23.32 € - 229.14 €
Chemical Identifiers
CAS | 2105-94-4 |
---|---|
Molecular Formula | C6H4BrFO |
Molecular Weight (g/mol) | 191.00 |
MDL Number | MFCD00011722 |
InChI Key | RYVOZMPTISNBDB-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene |
PubChem CID | 2724981 |
IUPAC Name | 4-bromo-2-fluorophenol |
SMILES | OC1=CC=C(Br)C=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449732
|
Thermo Scientific Alfa Aesar
A10804.06 |
5 g |
23.32 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459732
|
Thermo Scientific Alfa Aesar
A10804.14 |
25 g |
68.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469732
|
Thermo Scientific Alfa Aesar
A10804.22 |
100 g |
229.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2105-94-4 | |
191.00 | |
RYVOZMPTISNBDB-UHFFFAOYSA-N | |
2724981 | |
OC1=CC=C(Br)C=C1F |
C6H4BrFO | |
MFCD00011722 | |
2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene | |
4-bromo-2-fluorophenol |
Specifications
2105-94-4 | |
82°C to 83°C (10 mmHg) | |
C6H4BrFO | |
MFCD00011722 | |
2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene | |
OC1=CC=C(Br)C=C1F | |
191.00 | |
191 | |
4-Bromo-2-fluorophenol |
1.744 | |
98°C (208°F) | |
1.566 | |
1861281 | |
RYVOZMPTISNBDB-UHFFFAOYSA-N | |
4-bromo-2-fluorophenol | |
2724981 | |
≥97% |
Sicurezza e movimentazione
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.