missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorophenol, 97+%, Thermo Scientific Chemicals

Product Code. 11459732
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2105-94-4
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 191.00
MDL Number MFCD00011722
InChI Key RYVOZMPTISNBDB-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene
PubChem CID 2724981
IUPAC Name 4-bromo-2-fluorophenol
SMILES OC1=CC=C(Br)C=C1F

Specifications

Density 1.744
Boiling Point 82°C to 83°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.566
Quantity 25 g
Beilstein 1861281
Formula Weight 191
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-2-fluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.