Learn More
4-Bromo-2,3-difluorophenol, 97%, Thermo Scientific Chemicals
61.40 € - 205.00 €
Chemical Identifiers
CAS | 144292-32-0 |
---|---|
Molecular Formula | C6H3BrF2O |
Molecular Weight (g/mol) | 208.99 |
MDL Number | MFCD08061907 |
InChI Key | JZAVCMMYGSROJP-UHFFFAOYSA-N |
Synonym | 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol |
PubChem CID | 10219942 |
IUPAC Name | 4-bromo-2,3-difluorophenol |
SMILES | OC1=C(F)C(F)=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441290
|
Thermo Scientific Alfa Aesar
H26369.03 |
1 g | N/A | ||||||
11451290
|
Thermo Scientific Alfa Aesar
H26369.06 |
5 g |
205.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromo-2,3-difluorophenol is used as medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2,3-difluorophenol is used as medical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
144292-32-0 | |
208.99 | |
JZAVCMMYGSROJP-UHFFFAOYSA-N | |
10219942 | |
OC1=C(F)C(F)=C(Br)C=C1 |
C6H3BrF2O | |
MFCD08061907 | |
2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol | |
4-bromo-2,3-difluorophenol |
Specifications
144292-32-0 | |
MFCD08061907 | |
JZAVCMMYGSROJP-UHFFFAOYSA-N | |
4-bromo-2,3-difluorophenol | |
10219942 | |
97% |
C6H3BrF2O | |
2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol | |
OC1=C(F)C(F)=C(Br)C=C1 | |
208.99 | |
208.99 | |
4-Bromo-2,3-difluorophenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only