missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3-difluorophenol, 97%, Thermo Scientific Chemicals

Product Code. 11441290
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Bromo-2,3-difluorophenol is used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorophenol is used as medical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144292-32-0
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD08061907
InChI Key JZAVCMMYGSROJP-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol
PubChem CID 10219942
IUPAC Name 4-bromo-2,3-difluorophenol
SMILES OC1=C(F)C(F)=C(Br)C=C1

Specifications

Quantity 1 g
Formula Weight 208.99
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,3-difluorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'