missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzaldehyde, 97%, Thermo Scientific Chemicals

51.46 € - 621.00 €

Chemical Identifiers

CAS 40663-68-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00014133
InChI Key TYNJQOJWNMZQFZ-UHFFFAOYSA-N
Synonym 4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
PubChem CID 95942
IUPAC Name 4-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=C(C=O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404834
View Documents
Thermo Scientific Alfa Aesar
A13773.09
10 g
51.46 €
10g
Please to purchase this item. Need a web account? Register with us today!
11414834
View Documents
Thermo Scientific Alfa Aesar
A13773.18
50 g
158.79 €
50g
Please to purchase this item. Need a web account? Register with us today!
11424834
View Documents
Thermo Scientific Alfa Aesar
A13773.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
Specifications

Chemical Identifiers

40663-68-1
162.19
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
95942
C=CCOC1=CC=C(C=O)C=C1
C10H10O2
MFCD00014133
4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
4-prop-2-enoxybenzaldehyde

Specifications

40663-68-1
150°C to 152°C (18 mmHg)
C10H10O2
MFCD00014133
Air Sensitive
Immiscible in water.
C=CCOC1=CC=C(C=O)C=C1
162.19
162.19
4-Allyloxybenzaldehyde
1.058
>110°C (230°F)
1.5695
2206924
4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
4-prop-2-enoxybenzaldehyde
95942
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

missing translation for 'einecsNumber' : 255-027-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only