missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11424834
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40663-68-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00014133
InChI Key TYNJQOJWNMZQFZ-UHFFFAOYSA-N
Synonym 4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
PubChem CID 95942
IUPAC Name 4-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=C(C=O)C=C1

Specifications

Density 1.058
Boiling Point 150°C to 152°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5695
Quantity 250 g
Beilstein 2206924
Sensitivity Air Sensitive
Solubility Information Immiscible in water.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Allyloxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'