Learn More
4,5-Dichloro-o-phenylenediamine, 98%, Thermo Scientific Chemicals
70.00 € - 1020.00 €
Chemical Identifiers
CAS | 5348-42-5 |
---|---|
Molecular Formula | C6H6Cl2N2 |
Molecular Weight (g/mol) | 177.03 |
MDL Number | MFCD00007723 |
InChI Key | IWFHBRFJOHTIPU-UHFFFAOYSA-N |
Synonym | 4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine |
PubChem CID | 79297 |
IUPAC Name | 4,5-dichlorobenzene-1,2-diamine |
SMILES | NC1=CC(Cl)=C(Cl)C=C1N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493328
|
Thermo Scientific Alfa Aesar
A17738.06 |
5 g |
70.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403338
|
Thermo Scientific Alfa Aesar
A17738.14 |
25 g |
274.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413338
|
Thermo Scientific Alfa Aesar
A17738.22 |
100 g |
1020.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Chemical Identifiers
5348-42-5 | |
177.03 | |
IWFHBRFJOHTIPU-UHFFFAOYSA-N | |
79297 | |
NC1=CC(Cl)=C(Cl)C=C1N |
C6H6Cl2N2 | |
MFCD00007723 | |
4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine | |
4,5-dichlorobenzene-1,2-diamine |
Specifications
5348-42-5 | |
C6H6Cl2N2 | |
UN2811 | |
4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine | |
IWFHBRFJOHTIPU-UHFFFAOYSA-N | |
4,5-dichlorobenzene-1,2-diamine | |
79297 | |
98% |
∼162°C (decomposition) | |
MFCD00007723 | |
1072811 | |
Soluble in methanol and Dimethyl sulfoxide. Insoluble in water. | |
NC1=CC(Cl)=C(Cl)C=C1N | |
177.03 | |
177.03 | |
4,5-Dichloro-o-phenylenediamine |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 226-305-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only