missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Product Code. 11413338
Click to view available options
Quantity:
5 g
25 g
100 g

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5348-42-5
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00007723
InChI Key IWFHBRFJOHTIPU-UHFFFAOYSA-N
Synonym 4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine
PubChem CID 79297
IUPAC Name 4,5-dichlorobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(Cl)C=C1N

Specifications

Melting Point ∼162°C (decomposition)
Quantity 100 g
UN Number UN2811
Beilstein 1072811
Solubility Information Soluble in methanol and Dimethyl sulfoxide. Insoluble in water.
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 4,5-Dichloro-o-phenylenediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'