Learn More
4,4-Difluorocyclohexanone, 97%, Thermo Scientific Chemicals
170.00 € - 498.00 €
Chemical Identifiers
CAS | 22515-18-0 |
---|---|
Molecular Formula | C6H8F2O |
Molecular Weight (g/mol) | 134.126 |
MDL Number | MFCD08146635 |
InChI Key | NYYSPVRERVXMLJ-UHFFFAOYSA-N |
Synonym | 4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane |
PubChem CID | 15789092 |
IUPAC Name | 4,4-difluorocyclohexan-1-one |
SMILES | C1CC(CCC1=O)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15413347
|
Thermo Scientific Alfa Aesar
H26027.MD |
250 mg |
170.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11335055
|
Thermo Scientific Alfa Aesar
H26027.03 |
1 g |
498.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Chemical Identifiers
22515-18-0 | |
134.126 | |
NYYSPVRERVXMLJ-UHFFFAOYSA-N | |
15789092 | |
C1CC(CCC1=O)(F)F |
C6H8F2O | |
MFCD08146635 | |
4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane | |
4,4-difluorocyclohexan-1-one |
Specifications
22515-18-0 | |
C6H8F2O | |
4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane | |
NYYSPVRERVXMLJ-UHFFFAOYSA-N | |
4,4-difluorocyclohexan-1-one | |
15789092 | |
97% |
34°C to 40°C | |
MFCD08146635 | |
Insoluble in water. | |
C1CC(CCC1=O)(F)F | |
134.126 | |
134.12 | |
4,4-Difluorocyclohexanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only