missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorocyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 11335055
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Difluorocyclohexanone is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22515-18-0
Molecular Formula C6H8F2O
Molecular Weight (g/mol) 134.126
MDL Number MFCD08146635
InChI Key NYYSPVRERVXMLJ-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexanone, 4,4-difluoro-cyclohexanone, cyclohexanone, 4,4-difluoro, difluorocyclohexanone, acmc-1cnv9, 4,4-difluoro cyclohexanone, 4,4-difluoro-1-cyclohexanone, 4,4-difluorocycloxanone, 1,1,-difluoro-4-oxocyclohexane
PubChem CID 15789092
IUPAC Name 4,4-difluorocyclohexan-1-one
SMILES C1CC(CCC1=O)(F)F

Specifications

Melting Point 34°C to 40°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 134.12
Percent Purity 97%
Chemical Name or Material 4,4-Difluorocyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.