missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)pyridine, 99+%, Thermo Scientific Chemicals

53.00 € - 789.00 €

Chemical Identifiers

CAS 14548-48-2
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.652
MDL Number MFCD00006431
InChI Key YMTFKKLFLLAFNI-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
PubChem CID 98503
IUPAC Name (4-chlorophenyl)-pyridin-4-ylmethanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11465117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22551.03
1 g
53.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11475117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22551.06
5 g
202.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11485117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22551.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

14548-48-2
217.652
YMTFKKLFLLAFNI-UHFFFAOYSA-N
98503
C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl
C12H8ClNO
MFCD00006431
4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
(4-chlorophenyl)-pyridin-4-ylmethanone

Specifications

14548-48-2
C12H8ClNO
4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl
217.652
217.66
4-(4-Chlorobenzoyl)pyridine
107°C to 109°C
MFCD00006431
YMTFKKLFLLAFNI-UHFFFAOYSA-N
(4-chlorophenyl)-pyridin-4-ylmethanone
98503
≥99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 238-587-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only