missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorobenzoyl)pyridine, 99+%, Thermo Scientific Chemicals

Product Code. 11485117
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Chlorobenzoyl)pyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14548-48-2
Molecular Formula C12H8ClNO
Molecular Weight (g/mol) 217.652
MDL Number MFCD00006431
InChI Key YMTFKKLFLLAFNI-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl pyridine, 4-chlorophenyl pyridin-4-yl methanone, 4-4'-chlorobenzoyl pyridine, 4-chlorophenyl-pyridin-4-ylmethanone, methanone, 4-chlorophenyl-4-pyridinyl, acmc-1bu0c, 4-4-chloro-benzoyl pyridine, 4-chlorophenyl 4-pyridyl ketone, 4-chlorophenyl pyridin-4-ylmethanone, 4-chlorophenyl-4-pyridyl methanone
PubChem CID 98503
IUPAC Name (4-chlorophenyl)-pyridin-4-ylmethanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=NC=C2)Cl

Specifications

Melting Point 107°C to 109°C
Quantity 25 g
Formula Weight 217.66
Percent Purity ≥99%
Chemical Name or Material 4-(4-Chlorobenzoyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'