Learn More
4-(3-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals
73.50 € - 266.00 €
Chemical Identifiers
CAS | 63843-46-9 |
---|---|
Molecular Formula | C12H17NO |
Molecular Weight (g/mol) | 191.274 |
MDL Number | MFCD06248345 |
InChI Key | RTSXWJCYCFVWBU-UHFFFAOYSA-N |
Synonym | 4-3-methylphenoxy piperidine, 4-m-tolyloxy piperidine, acmc-1b64o |
PubChem CID | 17196730 |
IUPAC Name | 4-(3-methylphenoxy)piperidine |
SMILES | CC1=CC(=CC=C1)OC2CCNCC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499690
|
Thermo Scientific Alfa Aesar
H50950.MD |
250 mg | |||||||
11409700
|
Thermo Scientific Alfa Aesar
H50950.03 |
1 g |
266.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
63843-46-9 | |
191.274 | |
RTSXWJCYCFVWBU-UHFFFAOYSA-N | |
17196730 | |
CC1=CC(=CC=C1)OC2CCNCC2 |
C12H17NO | |
MFCD06248345 | |
4-3-methylphenoxy piperidine, 4-m-tolyloxy piperidine, acmc-1b64o | |
4-(3-methylphenoxy)piperidine |
Specifications
63843-46-9 | |
C12H17NO | |
4-3-methylphenoxy piperidine, 4-m-tolyloxy piperidine, acmc-1b64o | |
CC1=CC(=CC=C1)OC2CCNCC2 | |
191.274 | |
191.28 | |
4-(3-Methylphenoxy)piperidine |
190°C to 191°C | |
MFCD06248345 | |
RTSXWJCYCFVWBU-UHFFFAOYSA-N | |
4-(3-methylphenoxy)piperidine | |
17196730 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only